3-Chloro-4-fluorobenzamide 97%

Warning

A14392-03 A14392-06 A14392-03 A14392-06
AAA14392-06EA 180.93 USD
CAAAA14392-06 CAAAA14392-03 AAA14392-06 AAA14392-03
3-Chloro-4-fluorobenzamide 97%
3-Chloro-4-fluorobenzamide
Formula: C₇H₅ClFNO
Melting Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00153092
CAS Number: 701-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR