3-Chloro-4-fluorophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

A14382-06 A14382-06
AAA14382-06EA 35.49 USD
AAA14382-06 CAAAA14382-06
3-Chloro-4-fluorophenol 97%
3-Chloro-4-fluorophenol
Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 103…104 °C/11 mm
Melting Pt: 38…43 °C
Flash Pt: 108 °C(226 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002257
CAS Number: 2613-23-2
EINECS: 220-042-5
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR