4-Isopropoxybenzoic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

A14351-30 A14351-09 A14351-18 A14351-18 A14351-09 A14351-30
AAA14351-30EA 735.99 USD
CAAAA14351-30 AAA14351-30 AAA14351-18 AAA14351-09 CAAAA14351-18 CAAAA14351-09
4-Isopropoxybenzoic acid 99%
4-Isopropoxybenzoic acid
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 163…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00044318
CAS Number: 13205-46-4
EINECS: 236-169-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR