4'-Bromopropiophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: p-Bromopropiophenone

A14349-30 A14349-09 A14349-18 A14349-14 A14349-18 A14349-14 A14349-30 A14349-09
AAA14349-09EA 35.89 USD
CAAAA14349-30 AAA14349-09 AAA14349-18 AAA14349-30 AAAA14349-14 CAAAAA14349-14 CAAAA14349-18 CAAAA14349-09
4'-Bromopropiophenone 97%
4'-Bromopropiophenone
Formula: C₉H₉BrO
MW: 213.07 g/mol
Boiling Pt: 129…131 °C/7 mm
Melting Pt: 45…47 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000106
CAS Number: 10342-83-3
EINECS: 233-745-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR