3-Iodo-p-toluic acid 98%

Warning

Synonyms: 3-Iodo-4-methylbenzoic acid

A14197-14 A14197-06 A14197-14 A14197-06 A14197-22
AAA14197-06EA 40.46 USD
CAAAA14197-22 CAAAA14197-14 AAA14197-06 AAA14197-14 CAAAA14197-06
3-Iodo-p-toluic acid 98%
3-Iodo-p-toluic acid
Formula: C₈H₇IO₂
MW: 262.05 g/mol
Boiling Pt: 329 °C (1013 hPa)
Melting Pt: 206…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00010392
CAS Number: 82998-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR