2-Chloro-1,4-difluorobenzene 98%

Danger

Synonyms: 2,5-Difluorochlorobenzene

A14178-06 A14178-14 A14178-22 A14178-14 A14178-22 A14178-06
AAAA14178-22EA 376.35 USD
AAAA14178-22 AAA14178-14 CAAAAA14178-22 AAA14178-06 CAAAA14178-14 CAAAA14178-06
2-Chloro-1,4-difluorobenzene 98%
2-Chloro-1,4-difluorobenzene
Formula: C₆H₃ClF₂
MW: 148.54 g/mol
Boiling Pt: 126…128 °C
Melting Pt: –24.6 °C
Density: 1.352
Flash Pt: 20 °C(68 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042571
CAS Number: 2367-91-1
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR