3-Methylpyridazine 99%

Warning

A14097-06 A14097-22 A14097-14 A14097-22 A14097-14
AAA14097-22EA 1750.49 USD
CAAAA14097-22 CAAAA14097-14 AAA14097-22 CAAAA14097-06 AAA14097-14
3-Methylpyridazine 99%
3-Methylpyridazine
Formula: C₅H₆N₂
MW: 94.12 g/mol
Boiling Pt: 213…214 °C
Density: 1,030
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006469
CAS Number: 1632-76-4
EINECS: 216-640-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR