3,4-Difluorobenzaldehyde 98%

Warning

A14028-06 A14028-06 A14028-14 A14028-14
AAA14028-14EA 171.11 USD
AAA14028-14 AAA14028-06 CAAAA14028-14 CAAAA14028-06
3,4-Difluorobenzaldehyde 98%
3,4-Difluorobenzaldehyde
Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 180 °C
Density: 1.288
Flash Pt: 69 °C(156 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010328
CAS Number: 34036-07-2
EINECS: 422-180-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR