4'-Bromo-3-chloropropiophenone 94%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4'-Bromo-β-chloropropiophenone

A13988-18 A13988-09 A13988-06 A13988-06 A13988-18 A13988-09 A13988-30 A13988-30
AAAA13988-06EA 30.15 USD
AAAA13988-06 CAAAA13988-30 AAA13988-09 CAAAA13988-09 AAA13988-30 CAAAA13988-18 CAAAAA13988-06 AAA13988-18
4'-Bromo-3-chloropropiophenone 94%
4'-Bromo-3-chloropropiophenone
Formula: C₉H₈BrClO
MW: 247.52 g/mol
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 57…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00018979
CAS Number: 31736-73-9
EINECS: 250-784-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR