2,4-Dimethoxyaniline 97%
|
Warning
|
A13943-18
A13943-18
A13943-30
A13943-30
AAA13943-30EA
168.37
USD
AAA13943-30
AAA13943-18
CAAAA13943-30
CAAAA13943-18
2,4-Dimethoxyaniline 97%
2,4-Dimethoxyaniline
Formula:
C₈H₁₁NO₂ MW: 153.18 g/mol Melting Pt: 32…37 °C Density: 1.075 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008371 CAS Number: 2735-04-8 EINECS: 220-355-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...