2-(Trifluoromethoxy)benzamide 98%
|
Warning
|
A13889-06
A13889-06
A13889-03
A13889-03
AAA13889-06EA
184.26
USD
AAA13889-06
AAA13889-03
CAAAA13889-06
CAAAA13889-03
2-(Trifluoromethoxy)benzamide 98%
2-(Trifluoromethoxy)benzamide
Formula:
C₈H₆F₃NO₂ MW: 205.14 g/mol Melting Pt: 153…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042407 CAS Number: 127979-74-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...