3,4-Dichlorobenzamide 97%

A13878-06 A13878-03 A13878-03 A13878-06
AAA13878-06EA 245.12 USD
AAA13878-06 AAA13878-03 CAAAA13878-03 CAAAA13878-06
3,4-Dichlorobenzamide 97%
3,4-Dichlorobenzamide
Formula: C₇H₅Cl₂NO
Melting Pt: 140…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00053009
CAS Number: 2670-38-4
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR