Pentachloropyridine 98%

Warning

Synonyms: Perchloropyridine 2,3,4,5,6-Pentachloropyridine

A13842-36 A13842-0E A13842-0E A13842-36
AAA13842-0EEA 879.54 USD
CAAAA13842-0E CAAAA13842-36 AAA13842-0E AAA13842-36
Pentachloropyridine 98%
Pentachloropyridine
Formula: C₅Cl₅N
MW: 251.33 g/mol
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00006230
CAS Number: 2176-62-7
EINECS: 218-535-5
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR