trans-2,4-Dichlorocinnamic acid 98%

Warning

Synonyms: (2E)-3-(2,4-Dichlorophenyl)acrylic acid

A13824-22 A13824-06 A13824-14 A13824-14 A13824-22 A13824-06
AAAA13824-22EA 308.83 USD
CAAAAA13824-22 CAAAA13824-06 AAAA13824-22 CAAAA13824-14 AAA13824-06 AAA13824-14
trans-2,4-Dichlorocinnamic acid 98%
trans-2,4-Dichlorocinnamic acid
Formula: C₉H₆Cl₂O₂
MW: 217.05 g/mol
Melting Pt: 233…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00004373
CAS Number: 20595-45-3
EINECS: 000-000-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR