2-Methoxypyridine 98%

Supplier: Thermo Scientific Chemicals

Warning

A13675-36 A13675-14 A13675-14 A13675-36
AAA13675-14EA 36.31 USD
CAAAA13675-36 CAAAA13675-14 AAA13675-14 AAA13675-36
2-Methoxypyridine 98%
2-Methoxypyridine
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 142…143 °C
Density: 1.046
Flash Pt: 32 °C(89 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006262
CAS Number: 1628-89-3
EINECS: 216-623-8
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR