2-Octylthiophene 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-n-Octylthiophene

A13672-06 A13672-18 A13672-18 A13672-14 A13672-14 A13672-06
AAA13672-14EA 272.99 USD
AAA13672-14 AAA13672-06 AAAA13672-18 CAAAA13672-14 CAAAAA13672-18 CAAAA13672-06
2-Octylthiophene 98%
2-Octylthiophene
Formula: C₁₂H₂₀S
MW: 196.36 g/mol
Boiling Pt: 127…129 °C/12 mm
Density: 0.921
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041015
CAS Number: 880-36-4
EINECS: 212-913-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR