Cyclobutylamine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aminocyclobutane
A13423-03
A13423-14
A13423-06
A13423-03
A13423-14
A13423-06
AAA13423-06EA
197.99
USD
AAA13423-06
CAAAA13423-03
CAAAA13423-14
CAAAA13423-06
AAA13423-03
AAA13423-14
Cyclobutylamine 98%
Cyclobutylamine
Formula:
C₄H₉N MW: 71.12 g/mol Boiling Pt: 82…83 °C Density: 0.836 Flash Pt: -4 °C(24 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001328 CAS Number: 2516-34-9 EINECS: 219-736-0 UN: 2733 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...