3,4-Dibromotoluene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,2-Dibromo-4-methylbenzene
A13351-14
A13351-14
A13351-22
A13351-22
A13351-06
A13351-06
AAA13351-06EA
43.19
USD
AAA13351-06
AAA13351-14
CAAAA13351-06
AAA13351-22
CAAAA13351-14
CAAAA13351-22
3,4-Dibromotoluene 98+%
3,4-Dibromotoluene
Formula:
C₇H₆Br₂ MW: 249.93 g/mol Boiling Pt: 251 °C Melting Pt: –10 °C Density: 1,850 Flash Pt: 116 °C(241 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00079744 CAS Number: 60956-23-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...