2,4,5-Trifluorobenzonitrile 98+%

Warning

A13332-14 A13332-03 A13332-06 A13332-06 A13332-03 A13332-14
AAAA13332-14EA 295.53 USD
CAAAA13332-03 AAAA13332-14 CAAAA13332-06 CAAAAA13332-14 AAA13332-03 AAA13332-06
2,4,5-Trifluorobenzonitrile 98+%
2,4,5-Trifluorobenzonitrile
Formula: C₇H₂F₃N
MW: 157.09 g/mol
Boiling Pt: 168…170 °C
Density: 1.373
Flash Pt: 83 °C(181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013289
CAS Number: 98349-22-5
UN: 3276
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR