2,3-Diethylpyrazine 98%

Supplier: Thermo Scientific Chemicals
Warning

A13277-06 A13277-06 A13277-14 A13277-14 A13277-22 A13277-22
AAA13277-14EA 87.77 USD
AAA13277-14 CAAAA13277-06 CAAAA13277-14 AAA13277-06 CAAAA13277-22 AAA13277-22
2,3-Diethylpyrazine 98%
2,3-Diethylpyrazine
Formula: C₈H₁₂N₂
Boiling Pt: 180…182 °C
Density: 0.963
Flash Pt: 71 °C(160 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006151
CAS Number: 15707-24-1
EINECS: 239-800-1
ADR: 9,n/a

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR