Triphenylethylene 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1,2-Triphenylethylene1,1,2-Triphenylethene

A13244-09 A13244-18 A13244-30 A13244-30 A13244-18 A13244-09
AAA13244-09EA 68.29 USD
AAA13244-09 CAAAA13244-09 AAA13244-18 CAAAA13244-30 CAAAA13244-18 AAA13244-30
Triphenylethylene 98+%
Triphenylethylene
Formula: (C₆H₅)₂CCHC₆H₅
MW: 256.35 g/mol
Boiling Pt: 219…221 °C/14 mm
Melting Pt: 67…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00004765
CAS Number: 58-72-0
EINECS: 200-395-1
https://us.vwr.com/stibo/web/std.lang.all/07/00/760700.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR