3,4-Dichlorobenzyl alcohol 98%

Warning

Synonyms: (3,4-Dichlorophenyl)methanol

A13236-09 A13236-30 A13236-18 A13236-18 A13236-30 A13236-09
AAAA13236-30EA 548.9 USD
CAAAAA13236-30 AAAA13236-30 CAAAA13236-09 AAA13236-18 CAAAA13236-18 AAA13236-09
3,4-Dichlorobenzyl alcohol 98%
3,4-Dichlorobenzyl alcohol
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 130 °C/3 mm
Melting Pt: 36…39 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004633
CAS Number: 1805-32-9
EINECS: 217-299-0
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR