2-Fluoro-4-nitrophenol 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13134-14
A13134-06
A13134-06
A13134-14
A13134-03
A13134-03
AAA13134-14EA
304.99
USD
AAA13134-14
AAA13134-03
AAA13134-06
CAAAA13134-14
CAAAA13134-03
CAAAA13134-06
2-Fluoro-4-nitrophenol 98+%
2-Fluoro-4-nitrophenol
Formula:
C₆H₄FNO₃ MW: 157.1 g/mol Melting Pt: 117…121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051970 CAS Number: 403-19-0 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...