Tetrabromothiophene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Perbromothiophene

A13009-22 A13009-06 A13009-14 A13009-22 A13009-06 A13009-14
AAA13009-22EA 301.99 USD
AAA13009-22 CAAAA13009-22 CAAAA13009-06 CAAAA13009-14 AAA13009-06 AAA13009-14
Tetrabromothiophene 99%
Tetrabromothiophene
Formula: C₄Br₄S
MW: 399.73 g/mol
Boiling Pt: 324…327 °C
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00005419
CAS Number: 3958-03-0
EINECS: 223-554-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR