3-Fluoro-p-toluic acid 97%

Warning

Synonyms: 3-Fluoro-4-methylbenzoic acid

A12981-14 A12981-06 A12981-03 A12981-03 A12981-14 A12981-06
AAA12981-03EA 33.85 USD
AAA12981-03 CAAAAA12981-14 AAA12981-06 CAAAA12981-06 AAAA12981-14 CAAAA12981-03
3-Fluoro-p-toluic acid 97%
3-Fluoro-p-toluic acid
Formula: C₈H₇FO₂
MW: 154.14 g/mol
Boiling Pt: 263 °C (1013 hPa)
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00002490
CAS Number: 350-28-7
EINECS: 206-498-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR