Thiobenzamide 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Benzothioamide

A12973-22 A12973-14 A12973-36 A12973-14 A12973-36 A12973-22
AAA12973-22EA 162.99 USD
AAA12973-22 AAA12973-14 AAA12973-36 CAAAA12973-22 CAAAA12973-14 CAAAA12973-36
Thiobenzamide 98%
Thiobenzamide
Formula: C₇H₇NS
MW: 137.21 g/mol
Melting Pt: 114…118 °C
Flash Pt: 120 °C(248 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008060
CAS Number: 2227-79-4
EINECS: 218-765-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR