(2,4-Dichlorophenyl)acetonitrile 98+%

Warning

Synonyms: 2,4-Dichlorobenzyl cyanide

A12873-14 A12873-22 A12873-14 A12873-22
AAA12873-14EA 35.1 USD
CAAAA12873-22 AAA12873-14 CAAAA12873-14 AAA12873-22
(2,4-Dichlorophenyl)acetonitrile 98+%
(2,4-Dichlorophenyl)acetonitrile
Formula: C₈H₅Cl₂N
MW: 186.04 g/mol
Melting Pt: 58…62 °C
Flash Pt: 176 °C(348 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001899
CAS Number: 6306-60-1
EINECS: 228-621-4
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR