5-Bromovanillin 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Bromo-4-hydroxy-5-methoxybenzaldehyde

A12861-36 A12861-14 A12861-22 A12861-36 A12861-14 A12861-22
AAA12861-14EA 76.79 USD
CAAAA12861-22 AAA12861-14 AAA12861-22 AAAA12861-36 CAAAA12861-14 CAAAAA12861-36
5-Bromovanillin 98%
5-Bromovanillin
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 162…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00006940
CAS Number: 2973-76-4
EINECS: 221-016-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR