2-Hexylpyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

A12772-06 A12772-14 A12772-14 A12772-06
AAA12772-14EA 609.76 USD
AAA12772-14 CAAAA12772-06 AAA12772-06 CAAAA12772-14
2-Hexylpyridine 98%
2-Hexylpyridine
Formula: C₁₁H₁₇N
MW: 163.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00023552
CAS Number: 1129-69-7
EINECS: 214-454-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR