2-Ethylpyrazine 99%

Warning

Synonyms: Ethylpyrazine

A12759-06 A12759-06 A12759-14 A12759-14
AAA12759-06EA 52.46 USD
AAA12759-06 CAAAA12759-06 CAAAA12759-14 AAA12759-14
2-Ethylpyrazine 99%
2-Ethylpyrazine
Formula: C₆H₈N₂
Boiling Pt: 152…153 °C
Density: 0.988
Flash Pt: 42 °C(107 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006149
CAS Number: 13925-00-3
EINECS: 237-691-5
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR