Cyclohexylacetone 96%

Supplier: Thermo Scientific Chemicals

A12748-14 A12748-14 A12748-06 A12748-06
AAA12748-06EA 230.99 USD
AAA12748-06 CAAAA12748-06 CAAAA12748-14 AAA12748-14
Cyclohexylacetone 96%
Cyclohexylacetone
Formula: C₉H₁₆O
MW: 140.23 g/mol
Boiling Pt: 198…200 °C
Density: 0.905
Storage Temperature: Ambient
MDL Number: MFCD00019396
CAS Number: 103-78-6
EINECS: 203-143-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR