3-Amino-p-toluic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Amino-4-methylbenzoic acid
A12674-22
A12674-06
A12674-14
A12674-14
A12674-22
A12674-06
AAA12674-22EA
276.09
USD
AAA12674-22
AAA12674-14
CAAAA12674-14
CAAAA12674-06
CAAAA12674-22
AAA12674-06
3-Amino-p-toluic acid 99%
3-Amino-p-toluic acid
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Melting Pt: 164…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007742 CAS Number: 2458-12-0 EINECS: 219-543-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...