1-(4-Chlorophenyl)-2-thiourea 98%

Danger

Synonyms: 4-Chlorophenylthiourea N-(4-Chlorophenyl)thiourea

A12651-14 A12651-14
AAA12651-14EA 138.75 USD
CAAAA12651-14 AAA12651-14
1-(4-Chlorophenyl)-2-thiourea 98%
1-(4-Chlorophenyl)-2-thiourea
Formula: (C₆H₄Cl)NC(S)NH₂
MW: 186.66 g/mol
Melting Pt: 177…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00022168
CAS Number: 3696-23-9
EINECS: 223-022-4
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR