1-Phenylpiperazine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Phenylpiperazine
A12610-22
A12610-14
A12610-22
A12610-36
A12610-36
A12610-14
AAA12610-14EA
43.48
USD
CAAAA12610-22
CAAAA12610-36
CAAAA12610-14
AAA12610-14
AAA12610-36
AAA12610-22
1-Phenylpiperazine 98+%
1-Phenylpiperazine
Formula:
C₁₀H₁₄N₂ MW: 162.23 g/mol Boiling Pt: 285…286 °C Melting Pt: 17…19 °C Density: 1.064 Flash Pt: 141 °C(286 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005957 CAS Number: 92-54-6 EINECS: 202-165-6 UN: 2927 ADR: "6.1",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...