4-Pentylbiphenyl 99%

Synonyms: 4-n-Pentylbiphenyl4-Amylbiphenyl

A12590-06 A12590-14 A12590-06 A12590-14
AAA12590-14EA 243.09 USD
CAAAA12590-14 CAAAA12590-06 AAA12590-14 AAA12590-06
4-Pentylbiphenyl 99%
4-Pentylbiphenyl
Formula: C₁₇H₂₀
MW: 224.35 g/mol
Boiling Pt: 166…167 °C/6 mm
Melting Pt: 9…11 °C
Density: 0,945
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009503
CAS Number: 7116-96-3
EINECS: 230-421-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR