2-Aminobenzophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Aminobenzophenone

A12580-18 A12580-30 A12580-09 A12580-30 A12580-18 A12580-09
AAA12580-30EA 536.99 USD
CAAAA12580-18 CAAAA12580-09 CAAAA12580-30 AAA12580-30 AAA12580-18 AAA12580-09
2-Aminobenzophenone 98%
2-Aminobenzophenone
Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Boiling Pt: 280 °C
Melting Pt: 105…108 °C
Density: 1.32 g/cm³ (20 °C)
Flash Pt: 240 °C(464 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007713
CAS Number: 2835-77-0
EINECS: 220-613-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR