4-(Trifluoromethoxy)benzamide 97%

Warning

A12548-06 A12548-03 A12548-06 A12548-03
AAA12548-03EA 115.11 USD
AAA12548-03 CAAAA12548-06 AAA12548-06 CAAAA12548-03
4-(Trifluoromethoxy)benzamide 97%
4-(Trifluoromethoxy)benzamide
Formula: C₈H₆F₃NO₂
MW: 205.14 g/mol
Melting Pt: 155…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00041384
CAS Number: 456-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR