1,3,5-Trimethyl-1H-pyrazole 98%

Warning

Synonyms: 1,3,5-Trimethyl-1H-pyrazole

A12504-14 A12504-14 A12504-06 A12504-06
AAA12504-06EA 83.23 USD
CAAAA12504-14 CAAAA12504-06 AAA12504-06 AAA12504-14
1,3,5-Trimethyl-1H-pyrazole 98%
1,3,5-Trimethyl-1H-pyrazole
Formula: C₆H₁₀N₂
Boiling Pt: 168…170 °C
Melting Pt: 33…38 °C
Flash Pt: 33 °C(91 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015536
CAS Number: 1072-91-9
UN: 1325
ADR: "4.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR