2-Amino-6-fluorobenzonitrile 99%

Warning

Synonyms: 2-Cyano-3-fluoroaniline

A12456-06 A12456-14 A12456-06 A12456-03 A12456-03 A12456-14
AAA12456-14EA 283.79 USD
CAAAA12456-03 CAAAA12456-14 AAA12456-14 AAA12456-03 AAA12456-06 CAAAA12456-06
2-Amino-6-fluorobenzonitrile 99%
2-Amino-6-fluorobenzonitrile
Formula: C₇H₅FN₂
MW: 136.13 g/mol
Melting Pt: 125…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00015477
CAS Number: 77326-36-4
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR