2,2-Dichlorocyclopropylbenzene 97%

Synonyms: 1,1-Dichloro-2-phenylcyclopropane(2,2-Dichlorocyclopropyl)benzene

A12279-22 A12279-22 A12279-14 A12279-14 A12279-06 A12279-06
AAA12279-14EA 190.04 USD
CAAAA12279-06 CAAAA12279-14 AAA12279-14 AAA12279-22 AAA12279-06 CAAAA12279-22
2,2-Dichlorocyclopropylbenzene 97%
2,2-Dichlorocyclopropylbenzene
Formula: C₉H₈Cl₂
MW: 187.07 g/mol
Boiling Pt: 105…106 °C/12 mm
Density: 1,208
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001272
CAS Number: 2415-80-7
EINECS: 219-326-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR