2,3-Dichloroanisole 97%

Warning

Synonyms: 1,2-Dichloro-3-methoxybenzene

A12257-18 A12257-18 A12257-06
AAA12257-18EA 181.28 USD
CAAAA12257-18 AAA12257-18 AAA12257-06
2,3-Dichloroanisole 97%
2,3-Dichloroanisole
Formula: C₇H₆Cl₂O
MW: 177.03 g/mol
Boiling Pt: 115…116 °C/12 mm
Melting Pt: 31…33 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000545
CAS Number: 1984-59-4
EINECS: 217-853-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR