Nonanophenone 97%

Supplier: Thermo Scientific Chemicals

A12238-18 A12238-09 A12238-18 A12238-09
AAA12238-09EA 90.89 USD
AAA12238-09 CAAAA12238-18 CAAAA12238-09 AAA12238-18
Nonanophenone 97%
Nonanophenone
Formula: C₆H₅C(O)(CH₂)₇CH₃
MW: 218.34 g/mol
Boiling Pt: 139…140 °C/2 mm
Melting Pt: 14 °C
Density: 0.940
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048965
CAS Number: 6008-36-2
EINECS: 227-861-7
https://us.vwr.com/stibo/web/std.lang.all/22/67/762267.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR