1-Bromo-4-heptylbenzene 97%

Supplier: Thermo Scientific Chemicals

Synonyms: (4-Bromophenyl)heptane

A12235-14 A12235-06 A12235-03 A12235-06 A12235-14 A12235-03
AAA12235-06EA 104.62 USD
AAA12235-06 CAAAA12235-06 CAAAA12235-14 CAAAA12235-03 AAA12235-03 AAA12235-14
1-Bromo-4-heptylbenzene 97%
1-Bromo-4-heptylbenzene
Formula: C₁₃H₁₉Br
MW: 255.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00061115
CAS Number: 76287-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR