4-Bromo-2-(trifluoromethyl)aniline 98%

Warning

Synonyms: 2-Amino-5-bromo-α,α,α-trifluorotoluene 2-Amino-5-bromo-α,α,α-trifluortoluene 4-Bromo-α,α,α-trifluoro-o-toluidine

A12146-14 A12146-06 A12146-14 A12146-06
AAA12146-14EA 156.56 USD
CAAAA12146-14 AAA12146-14 CAAAA12146-06 AAA12146-06
4-Bromo-2-(trifluoromethyl)aniline 98%
4-Bromo-2-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 84…85 °C/5 mm
Density: 1.710
Flash Pt: 107 °C(224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064393
CAS Number: 445-02-3
EINECS: 207-150-8
UN: 2810
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR