Perfluoroheptanoic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Tridecafluoroenanthic acid Tridecafluoroheptanoic acid Tridecafluoroheptylic acid Perfluoroenanthic acid

A12092-14 A12092-06 A12092-06 A12092-14
AAA12092-14EA 194.99 USD
AAA12092-14 AAA12092-06 CAAAA12092-14 CAAAA12092-06
Perfluoroheptanoic acid 98+%
Perfluoroheptanoic acid
Formula: C₇HF₁₃O₂
Boiling Pt: 89 °C/30 mm
Melting Pt: 31…36 °C (lit.)
Density: 1.792
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039604
CAS Number: 375-85-9
EINECS: 206-798-9
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR