4-Pentylbenzoic acid 99%

Warning

A12086-06 A12086-06 A12086-14 A12086-14
AAA12086-06EA 61.8 USD
CAAAA12086-14 AAA12086-06 AAA12086-14 CAAAA12086-06
4-Pentylbenzoic acid 99%
4-Pentylbenzoic acid
Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
Boiling Pt: 260,7 °C (1013 hPa)
Melting Pt: 85…88 °C
Density: ∼0,992 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002572
CAS Number: 26311-45-5
EINECS: 247-607-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR