2-Isopropylphenol 98%

Supplier: Thermo Scientific Chemicals
Danger

A12027-18 A12027-18
AAAA12027-18EA 41.29 USD
AAAA12027-18 CAAAAA12027-18
2-Isopropylphenol 98%
2-Isopropylphenol
Formula: C₉H₁₂O
MW: 136.19 g/mol
Boiling Pt: 212…214 °C
Melting Pt: 14…16 °C
Density: 1.010
Flash Pt: 88 °C(190 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002224
CAS Number: 88-69-7
EINECS: 201-852-8
UN: 2922
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR