1-Bromo-2,4,5-trifluorobenzene 98+%

Warning

Synonyms: 2,4,5-Trifluorobromobenzene

A11962-03 A11962-06 A11962-06 A11962-14 A11962-03 A11962-14
AAA11962-06EA 56.11 USD
CAAAA11962-03 CAAAA11962-14 AAA11962-06 AAA11962-03 AAA11962-14 CAAAA11962-06
1-Bromo-2,4,5-trifluorobenzene 98+%
1-Bromo-2,4,5-trifluorobenzene
Formula: C₆H₂BrF₃
MW: 210.98 g/mol
Boiling Pt: 143…144 °C
Melting Pt: –19 °C
Density: 1.806
Flash Pt: 55 °C(131 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000306
CAS Number: 327-52-6
EINECS: 206-318-8
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR