7-Methoxy-4-methylcoumarin 99%

Synonyms: 7-Methoxy-4-methyl-2-benzopyrone

A11785-06 A11785-03 A11785-06 A11785-03
AAA11785-03EA 75.1 USD
CAAAA11785-06 AAA11785-03 AAA11785-06 CAAAA11785-03
7-Methoxy-4-methylcoumarin 99%
7-Methoxy-4-methylcoumarin
Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
Boiling Pt: 130…144 °C (0,3 mmHg)
Melting Pt: 159…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00009773
CAS Number: 2555-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR