α,2-Dichloro-6-fluorotoluene 98%

Danger

Synonyms: 2-Chloro-6-fluorobenzyl chloride

A11758-22 A11758-14 A11758-22 A11758-14
AAA11758-22EA 100.87 USD
CAAAA11758-22 AAA11758-22 AAA11758-14 CAAAA11758-14
α,2-Dichloro-6-fluorotoluene 98%
α,2-Dichloro-6-fluorotoluene
Formula: C₇H₅Cl₂F
MW: 179.02 g/mol
Boiling Pt: 208…210 °C
Melting Pt: 7 °C
Density: 1.401
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000896
CAS Number: 55117-15-2
EINECS: 259-487-5
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR